Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHAPSO (White Powder), Fisher BioReagents

Catalog Number BP575500
Click to view available options
Quantity:
500 mg
Packaging:
Amber Glass
82473-24-3
C32H58N2O8S
630.882
GUQQBLRVXOUDTN-AKMBKZMMSA-N
3-[(3-Cholamidopropyl) dimethylammonio]-2-hydroxy-1-propanesulfonate
134129643
3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8S,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]-2-hydroxypropane-1-sulfonate
CC(CCC(=O)NCCC[N+](C)(C)CC(CS(=O)(=O)[O-])O)C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C
82473-24-3
C32H58N2O8S
630.882
GUQQBLRVXOUDTN-AKMBKZMMSA-N
3-[(3-Cholamidopropyl) dimethylammonio]-2-hydroxy-1-propanesulfonate
134129643
3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8S,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]-2-hydroxypropane-1-sulfonate
CC(CCC(=O)NCCC[N+](C)(C)CC(CS(=O)(=O)[O-])O)C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C
CHAPSO is a Zwitterionic detergent used for protein solubilization.
TRUSTED_SUSTAINABILITY
Absorbance 0.05 max. (1% solution in water) at 260nm
Color White
Physical Form Powder
Melting Point 184°C
Quantity 500 mg
Merck Index 15, 2043
Identification Pass Test
Formula Weight 630.88
Ignition Residue 0.1% max.
Packaging Amber Glass
Water 7.5% max.
Chemical Name or Material CHAPSO
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Harmful by inhalation, in contact with skin and if swallowed. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

NFPA

Health:2
Flammability:1
Instability:0
Recommended Storage RT
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit