Learn More
Clontech Labs 3P Chaperone Plasmid Set, 1 Set
Supplier: Clontech Labs 3P 3340
Consists of five different plasmids, each of which is designed to express multiple molecular chaperones that function as a "chaperone team" to enable protein folding. Co-expression of a target protein with one of these chaperone teams increases the recovery of soluble proteins. Each plasmid carries an origin of replication derived from pACYC and a Cmr gene, which allows use with E. coli expression systems utilizing ColE1-type plasmids containing an ampicillin resistance gene as a marker. The chaperone genes are situated downstream of an araB or Pzt-1 (tet) promoter. Therefore, expression of target proteins and chaperones can be induced individually if the target gene is placed under the control of other promoters (e.g. lac). These plasmids also contain the necessary regulator (araC or tetr) for each promoter. 1 Set
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.