Learn More
Electron Microscopy Sciences Chang Mold 10 Cavities, 2 cm x 2 cm
Supplier: Electron Microscopy Sciences 70920
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Designed by Dr. J.P. Chang for the embedding of a monolayer of cells. This specially designed mold has proven useful in the processing of frozen sections, ascite cells, fresh imprints, as well as blood and marrow smears.(J. of Ultrastructural Res., 37, 370 1971).
Size: 2 cm x 2 cm