Learn More
Darwin Chambers Company Double Door Plant Growth Chamber 3 Tiers
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Darwin Chambers Company PG0683
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
![Request Bulk Order](/content/dam/fishersci/glyphs/15395_Quote_Bulk_EN.jpg)
The PG068 is a 69.3 cubic feet double door plant growth chamber. Temperature range is 4°C to 60°C (lights off) 10°C to 60°C (lights on).Optional humidity range ambient to 90 percent. Humidity control can be added as a chargeable option. This unit comes with 6 stainless steel shelves. Lighting and lighting control are included in chamber. The chamber provides 304 series stainless steel interior and door liner, corrosion-resistant; also a heavy-gauge powder-coated, galvanized steel exterior. The PG Series chambers feature hot gas by-pass technology which controls the temperature of the epoxy coated evaporator for greater uniformity. The ultrasonic humidification system provides excellent humidity control and avoids hot spots seen during chamber mapping of steam boiler equipped chambers.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.