Learn More
Chemglass Life Sciences Vapor Tube, Compatible with Buchi Rotary Evaporators, 24/40, 188mm OAL
Supplier: Chemglass Life Sciences CG1317G20
Decaled "R210/215", for Glass Assembly (V,C,S,E,R,BY). Length "A" = 52mm, 188mm OAL. Replacement Vapor tubes compatible with Buchi rotary evaporators. Thread is made for use with standard Combi-Clip. NOT SUPPLIED WITH COMBI-CLIP. Buchi is a registered trade mark of BÜCHI Labortechnik AG.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.