Learn More
Chondrex Inc CFA, 10 MG/ML x 5 ML
Supplier: Chondrex Inc 7027
Heat-killed M. tuberculosis H37 RA (non-viable) suspended in Incomplete Freund's Adjuvant at a concentration of 10 mg/ml. 5 mL.