Learn More
Strem, An Ascensus Company CAS# [7440-46-2]. 1g. Cesium (99.9+%) (prescored ampoule). MFCD00134037
Supplier: Strem, An Ascensus Company 5555511G
CAS# 7440-46-2. 1g. Cesium (99.9+%) (prescored ampoule). MFCD00134037. Molecular Weight: 132.91. Molecular Formula: Cs. Color/form: under argon Strem# 55-5551. www.strem.com/catalog/v3/55-5551/