Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ceric Ammonium Nitrate, 0.05 N Solution, Spectrum™ Chemical
Specifications
CAS | 16774-21-3, 7732-18-5, 7697-37-2 |
---|---|
Concentration or Composition (by Analyte or Components) | 0.050 ± 0.001 N |
Packaging | Amber Glass Bottle |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
18-609-606
|
Spectrum Chemical Manufacturing Corporation
C146500MLGL |
500 mL | Amber Glass Bottle | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
18-609-607
|
Spectrum Chemical Manufacturing Corporation
C1464LTGL |
4 L | Amber Glass Bottle | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Specifications
16774-21-3, 7732-18-5, 7697-37-2 | |
0.050 ± 0.001 N | |
Amber Glass Bottle |
CAUTION: For laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title