Learn More
Hach Company Replacement ceramic junction for 5044 reference electrode
Supplier:  Hach Company LZW5243.99
Encompass_Preferred
LZW5243.99 is a replacement ceramic diaphragm (junction) for the 5044 reference electrode, recommended for use with Hach sensION+ ion selective electrodes (ISE).
Catalog No. 50-199-7982