Learn More
Cole-Parmer Raven Environmental F-10300 Benchtop Centrifuge; 230 VAC, 50 Hz
Supplier:  Cole-Parmer 0547715
Encompass
Heavy-duty stainless steel and aluminum construction makes this benchtop centrifuge ideal for industrial environments such as food and beverage, wastewater plants, meat packing, and paper mills. Includes six polycarbonate reusable tubes clearly marked to indicate percent volume. The conical shape of the tubes accentuate small amounts of solids concentration to as low as 0.2%. Separate solids from liquid for improved laboratory tests.
Catalog No. 50-632-417