Learn More
Eppendorf™ Centrifuge 5702 RH
Centrifuge 5702 RH - Quiet in operation, compact in design and provide six different rotor options to accommodate any type of tube and requirement for low to mid-throughput applications in clinical research and cell culture.
$9090.00 - $10190.00
Catalog Number | Mfr. No. | Includes | Rotor Included | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Rotor Included | Price | Quantity & Availability | ||||
05-414-123
|
Eppendorf™
022626213 |
Without Rotor | No |
Each for $9,090.00
|
|
||||
05-414-124
|
Eppendorf™
022628023 |
Adapters for 13/16 mm Blood Collection Tubes | Yes |
Each for $10,190.00
|
|
||||
05-414-126
|
Eppendorf™
022628124 |
Adapters for 15/50 mL Conical Tubes | Yes |
Each for $10,190.00
|
|
||||
Description
- Rotary Knobs, Refrigerated/Heated
- 120 V, 50/60 Hz (US)
Specifications
Centrifuge | |
120 V | |
50/60 Hz |