Learn More
Thermo Scientific CENTER TUBE
Supplier:  Thermo Scientific 842312051971
Encompass
Centre Tube 2mm ID (EMT Torch); Centre tube intended for samples containing high dissolved solids.; Supplied as standard on all Duo iCAP instruments and can only be used; with the centre tube holder for the EMT torch.
Catalog No. NC9758015