Learn More
Sigma Aldrich Fine Chemicals Biosciences Celpure P300, 91053-39-3, MFCD00132803, 1 kg
Supplier: Sigma Aldrich Fine Chemicals Biosciences 5252431KG
Meets USP/NF testing specifications, Synonym: Diatomaceous earth, calcined. Celpure P300 has been used used in the column chromatographic isolation and extraction of testosterone and corticosterone from freeze-dried water dragon plasma samples.