Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cellucap Spunbond Polypropylene Shoe Covers
$171.00 - $179.00
Specifications
Product Type Specs | Shoe Covers |
---|---|
Material | Polypropylene |
Color | Blue |
Resistance | Fluid |
For Use With (Application) | Ideal for visitors, light duty applications or general purpose coverage |
Specifications
Shoe Covers | |
Blue | |
Ideal for visitors, light duty applications or general purpose coverage |
Polypropylene | |
Fluid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title