Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cellucap 1.25 oz. Spunbond Polypropylene Lab Coats
$16.07 - $263.00
Specifications
Material | Polypropylene |
---|---|
Color | White |
Closure Type | Hook and Loop |
Disposable | Yes |
For Use With (Application) | Splash protection from nonhazardous liquids and dry particulates |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-304-131
|
Cellucap
3302EWHLS |
Small |
Case of 25 for $186.00
|
|
19-304-136
|
Cellucap
3302EWHLXXX |
3X-Large |
Case of 25 for $232.00
|
|
19-304-134
|
Cellucap
3302EWHLX |
X-Large |
Case of 25 for $186.00
|
|
19-304-132
|
Cellucap
3302EWHLM |
Medium |
Case of 25 for $175.50
|
|
19-304-135
|
Cellucap
3302EWHLXX |
2X-Large |
Case of 25
for $16.07
Save 93% Reg : |
|
19-304-137
|
Cellucap
3302EWHLXXXX |
4X-Large |
Case of 25 for $203.50
|
|
19-304-133
|
Cellucap
3302EWHLL |
Large |
Case of 25 for $186.00
|
|
19-304-139
|
Cellucap
3302EWHLXXXXXX |
6X-Large |
Case of 25
for $20.28
Save 93% Reg : |
|
19-304-138
|
Cellucap
3302EWHLXXXXX |
5X-Large |
Case of 25 for $230.00
|
|
19-304-140
|
Cellucap
3302EWHLXXXXXXX |
7X-Large |
Case of 25 for $263.00
|
|
Specifications
Polypropylene | |
Hook and Loop | |
Splash protection from nonhazardous liquids and dry particulates | |
0.03 kg |
White | |
Yes | |
1.25 oz. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title