Learn More
SLINGSHOT BIOSCIENCES CELLTRAX TOX FOR 50 TESTS
SDP
Supplier:  SLINGSHOT BIOSCIENCES SSB28A
Encompass
CellTraX™ Tox viability controls are advanced 2-in-1 beads that bind to DNA intercalating dyes (7AAD, PI) and amine-reactive viability dyes to simulate the staining of cells.This product is intended as a process and assay control to match the viability staining of real cells. Packaged in two bottles with the positive and negative controls in separate vials.
Catalog No. NC2598904