Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Greiner Bio-One CELLSTAR 384-well, Cell Culture-Treated, Flat-Bottom, Small Volume, High Base Microplate
Color:Black; Sterility:Sterile; Lid:With Lid
Supplier: Greiner Bio-One 784086
Description
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic Non-cytotoxicCompatible with automated systemsFlexible working volumeRounded square well profile
- Stackable
- Alphanumeric well cod
Specifications
No | |
With Lid | |
Black | |
384 | |
Cell Culture | |
Round, Flat Bottom (F-well) | |
8 |
Standard | |
16 x 24 | |
Polystyrene | |
Sterile | |
4 to 25 μL | |
32 | |
Black |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction