Learn More
Alkali Scientific CellPro™ Reducing Adapter for use with 15mL, 17x100mm, 12x75mm and 5mL conical tubes, pack of 25
Supplier: Alkali Scientific MT4200
Premium Cell Strainer system is a fast and easy-to-use solution for detaching, isolating and straining cells from clusters and tissues. A polypropylene frame supports a nylon mesh that is available in three color-coded porosities: 40μm (blue), 70μm (white), and 100μm (yellow). The advanced frame/housing design also contributes to superior performance and reduced processing time, by eliminating the “air-lock” effect. Improved aeration allows air to escape while filtrate is entering the tube, preventing messy spillovers.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.