Learn More
Cellpath Stainless-Steel Supa Mega Base Moulds
Reusable base moulds. CellPath Stainless-Steel Supa Mega Base Moulds have extra deep side walls allowing wax to be decanted into the cassette during embedding without the risk of the usual ‘leakage’ of wax.
$251.00
Catalog Number | Mfr. No. | Length (Metric) | Width (Metric) | Depth (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Width (Metric) | Depth (Metric) | Price | Quantity & Availability | |||
22-222-030
|
Cellpath
GBC601405A |
60 mm | 45 mm | 15mm |
Pack of 5 for $251.00
|
|
|||
22-222-031
|
Cellpath
GBC361405A |
36 mm | 36 mm | 10mm | N/A | N/A | |||
This product is either new or inactive, and cannot be ordered online.Inactive Item. Please call Customer Service Representative. | |||||||||
Description
- Reusable
- Stackable to reduce storage area
Specifications
5/Pk. |