Learn More
Cellink Inc CELLINK START (50 ML)
Supplier:  Cellink Inc IK1900005001
Encompass
CELLINK START is a water-soluble gel used as a sacrificial material for cell-laden constructs, bioinks with poor shape fidelity and constructs with porosity along all three axes.
Catalog No. NC2143471