Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One CELLCOAT™ Poly-D-lysine Coated Cell Culture Dish

For cultivation in serum-free and serum reduced medium

$615.00 - $1195.00

Specifications

Closure Color Clear
Closure Included Closure Included
Closure Material Polystyrene
Bottom Shape Flat
Color Clear
View More Specs

Products 2
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
07-000-189
View Documents
Greiner Bio-One
664940
100 mm
Case of 40 for $615.00
Add to Cart
 
07-000-587
View Documents
Greiner Bio-One
628940
60 mm
Case of 100 for $1,195.00
Add to Cart
 
Description

Description

Greiner Bio-One CELLCOAT Poly-D-lysine Coated Cell Culture Vessel is used for cultivation of transfected cells and fastidious cell lines, as well as serum-free or -reduced cell culture.

  • Source: Synthetic
  • Cultivation of neuronal cells, transfected cells and other fastidious cell lines
  • Cultivation in serum-free and serumreduced medium
  • Improvement of cell attachment
Specifications

Specifications

Clear
Polystyrene
Clear
Round
Poly-D-Lysine
Lid
Closure Included
Flat
Polystyrene
Non-sterile
Cell Culture Dish
SDS
Documents

Documents

Product Certifications

For research use only. Not for use in diagnostic procedures, unless otherwise indicated

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit