Learn More
Research Products International Corp Cell Scraper, Pivoting 1.8cm Blade, Sterile, Non-Pyrogenic, 18cm Long Handle, 100 per Case
SDP
Supplier:  Research Products International Corp 162422
Encompass_Preferred
Cell Scrapers feature 1.8cm blades for precise cutting and scraping of cells.
Catalog No. 50-476-477