Learn More
Alkali Scientific Cell Lifter, Double Blade with 23mm & 12mm, 218mm handle, sterile, 100/Case
Supplier: Alkali Scientific TC7023
• Useful for minimal harvesting of cells• 2 types of blades in handle, wide/narrow• Individually packed• Pyrogen Free• Sterile• QTY: 100/Case