Learn More
Thomas Scientific Cell Lifter, Double End, Flat Blade & J-Hook
Supplier: Thomas Scientific 1159T47
Qty 100 Individually Wrapped Double Ended Cell Lifters have a 19mm beveled edge blade on one end and a J-Hook or Narrow Blade on the other for removing cells from Multiple Well Plates or Tubes J-Hook is curved on the end to vertically lift cells from 6, 12, 24 and 48 Well Plates Narrow Blade is tapered to fit into a 96 Well Plate or Micro Centrifuge Tubes Cell Scrapers & Cell Lifters are non-pyrogenic and packaged sterile (gamma irradiated)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.