Learn More
Abbott Laboratories™ CELL-DYN Emerald
Compact hematology analyzer that delivers reliable, 3-part differentials for low-volume laboratories.
Supplier: Abbott Laboratories™ 09H3901
This item is not returnable.
View return policy
Description
- Stores up to 1,500 records with histograms on internal memory: up to 60,000 on external USB thumb drive
- Has easy to use software controlled by a color touchscreen with a built-in keyboard
- System utilizes electrical impedance for WBC, RBC, and platelet counting and absorption spectrophotometry for hemoglobin
- Package includes Cell-Dyn Emerald analyzer, UPS, and freight
- Reagents: 2 and cleaner
- Sampling: Open tube only
- Maintenance Requirements: 2 minute bleach clean cycle weekly, manual syringe lubrication annualy
Specifications
9.8 μL | |
13.8 in. | |
9.8 in. | |
16 |
57/hr | |
13.8 in. | |
19.8 lb. |