Learn More
BIOLOGIX USA INC CELL CULTR DISH 60X20 LOW BIND
Supplier: BIOLOGIX USA INC 911625
Weight: 0.88 lbs▪ Mesh membrane material: Nylon▪Frame material: PS▪ Mesh membrane thickness: 10 ~ 23 μm▪ Membrane pore size: 0.40 μm