Learn More
Perkin Elmer US LLC Absorption Cell Body for FIMS Instruments
Supplier: Perkin Elmer US LLC B0510370
Replacement absorption cell body for the Flow Injection Mercury System (FIMS). The FIMS-cell has an inner diameter of 4 mm and an optical path length of 260 mm.