Learn More
Celestron™ LCM Computerized Telescope

Automatically locates the wonders of the universe with its motorized system and on board computer.
Supplier: Celestron™ 31150
Includes: CD-ROM The SkyX Astronomy Software provides education about the sky and printable sky maps, sturdy aluminum tripod and accessory tray.
Description
Database allows telescope to locate over 4,000 celestial objects.
- Quick-release computerized base, optical tube and accessory tray for quick, no-tool set up
- Proven NexStar computer control technology
- SkyAlign allows to align on any three bright celestial objects, making for a fast and easy alignment process
- Flash upgradeable hand control software and motor control units for downloading product updates over the Internet
- Internal battery compartment to prevent cord wrap during use
- Compatible with optional NexRemote telescope control software, for advanced control of your telescope via computer
60LCM Computerized TelescopeHigh quality 60mm refractor
Specifications
Fully Coated | |
Computerized Telescope | |
40x, 111x | |
Motorized Altazimuth | |
Newtonian Reflector | |
Angular: 1.6°, linear: 84 in. |
100 cm | |
114 mm | |
114LCM | |
1.22 Arc sec. (Rayleigh), 1.02 arc sec. (Dawes) | |
39.37 in. | |
CD ROM The SkyX astronomy software provides education about the sky and printable sky maps, sturdy aluminum tripod and accessory tray |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.