Learn More
Mabtech Inc CEF peptide pool for human CD8 T cells
Supplier: Mabtech Inc 36161
The CEF peptide pool stimulates CD8+ T cells to produce preferentially IFN-γ and is recommended as a positive control in IFN-γ ELISpot and FluoroSpot assays using human PBMC. The CEF peptide pool consists of 23 MHC class I-restricted viral peptides from human CMV, EBV and influenza virus. It has been shown to induce IFN-γ responses in almost 90% of all Caucasians.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.