Learn More
Thermo Scientific™ CEDIA™ MultiDrug Drugs of Abuse Calibrators
Utilize the globally valued accuracy, precision and lot-to-lot dependability of CEDIA™ Multidrug Drugs of Abuse Calibrators.
$140.00
Specifications
Content And Storage | Do not Freeze; 2° to 8°C |
---|---|
Concentration | Multianalyte |
Detectable Analytes | Multiple |
Sample Type | Urine |
Type | Calibrator |
Catalog Number | Mfr. No. | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Quantity | Price | Quantity & Availability | ||||
23-278-258
|
Thermo Scientific™
1730380 |
CEDIA™ Intermediate Calibrator | 1 x 5 mL |
Each for $140.00
|
|
||||
23-278-257
|
Thermo Scientific™
1730398 |
CEDIA™ High Calibrator | 1 x 5 mL |
Each for $143.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-278-248
|
Thermo Scientific™
1730401 |
CEDIA™ Clinical Calibrator | 1 x 5 mL |
Each for $138.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-278-260
|
Thermo Scientific™
1730428 |
CEDIA™ Secondary Calibrator | 1 x 5 mL |
Each for $143.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-278-252
|
Thermo Scientific™
1730517 |
CEDIA™ Secondary Calibrator | 1 x 15 mL |
Each for $317.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-278-250
|
Thermo Scientific™
1732218 |
CEDIA™ Intermediate Calibrator | 1 x 15 mL |
Each for $335.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-278-256
|
Thermo Scientific™
1732226 |
CEDIA™ High Calibrator | 1 x 15 mL |
Each for $325.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-278-251
|
Thermo Scientific™
1815326 |
CEDIA™ Primary Calibrator | 1 x 5 mL |
Each for $138.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-278-246
|
Thermo Scientific™
1815334 |
CEDIA™ Primary Calibrator | 1 x 15 mL |
Each for $335.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Utilize the globally valued accuracy, precision and lot-to-lot dependability of CEDIA™ Multidrug Drugs of Abuse Calibrators, which are optimized for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing on the market.
- Qualitative or semi-quantitative options for results
- Excellent correlation to GC/MS
- Applications for an array of clinical chemistry analyzers
- Variety of calibrators and multiconstituent control sets
Specifications
Do not Freeze; 2° to 8°C | |
Multiple | |
Calibrator |
Multianalyte | |
Urine | |
1 Bottle |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.