Learn More
Thermo Scientific™ CEDIA™ Benzodiazepine Assays
Utilize the globally valued, highly accurate, and precise Thermo Scientific™ CEDIA™ Benzodiazepine Assay for very stable shelf life and lot-to-lot dependability.
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Control Sets | MGC Multi-Drug Controls |
Detectable Analytes | Nitrazepam |
DoA Calibrators | CEDIA Multi-drug Calibrators |
Sample Type | Urine |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-278-208
|
Thermo Scientific™
100085 |
3 x 17 mL, TSC |
Each for $1,687.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-278-206
|
Thermo Scientific™
10016409 |
3 x 17 mL, Indiko |
Each for $711.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-278-207
|
Thermo Scientific™
100094 |
1 x 65 mL, MCC |
Each for $1,867.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Utilize the globally valued, highly accurate, and precise Thermo Scientific™ CEDIA™ Benzodiazepine Assay for very stable shelf life and lot-to-lot dependability. Applications are available for an array of clinical chemistry analyzers. The β-Glucuronidase Enzyme, when used with this assay, offers enhanced sensitivity of parent drug by hydrolyzing the glucuronide metabolites. The Thermo Scientific CEDIA panel of drugs of abuse assays is ever expanding to address emerging drug trends and testing needs.
- One of the largest menus of tests available
- Qualitative or semi-quantitative options for results
- Excellent correlation to GC/MS
- Variety of calibrators and multiconstituent control sets
Specifications
2°C to 8°C | |
Nitrazepam | |
Urine |
MGC Multi-Drug Controls | |
CEDIA Multi-drug Calibrators | |
Reagent |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.