Learn More
Andwin Scientific CDEHL STR 70%IPA 16OZ TRG 12CS
Supplier: Andwin Scientific 89108160
The Sterile 70% Isopropanol is filtered to 0.2m and induction sealed to guarantee its sterility. This product is composed of 70% USP grade isopropanol and 30% USP grade deionized water. It is perfect for use with sterile cleanroom wipers. This is a