Learn More
Thermo Scientific Dionex ICS-6000 CD Conductivity Detector | | 1 EA
Supplier: Thermo Scientific 079829
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
High Sensitivity with Low Noise Across Concentrations, Excellent detection from high to low concentrations with microprocessor-controlled digital signal processing and wide dynamic range of up to 15000Scm, Improved stability and lower noise from the integrated electronics between the cell and detector, Enhanced thermal stability with independent detector temperature control, Choice of capillary and analytical standard and microbore models, Flexible configuration for use in series with another detector or parallel for dual detection, Simple snap-in installation no cables or tools required automatically recognized by Thermo Scientific Chromeleon Chromatography Data System software, Compact installation requiring minimal tubing lengths with up to two detector modules in the same ICS-5000ICS-6000 DC DetectorChromatography Compartment, Capillary CD Detector
Description
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.