Learn More
Hach Company Radiometer Analytical CDC861T Conductivity Cell with Temperature Sensor (4-pole Pt sensor,k = 1.0 cm-1, glass body)
Supplier: Hach Company E61M016
The Radiometer CDC861T is a 4-pole conductivity cell with platinized, platinum sensor (k = 1.0 cm-1), integrated temperature sensor and glass body. The CDC861T sensor's 4 platinum ring design and glass body ensures results in general aqueous and non-aqueous media, including very strong acids and bases or saline media, over several decades of conductivity with a single cell, using just one calibration. It has a fixed cable with MAB6 plug for use with Radiometer Analytical CDM210/230/92 conductivity meters.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.