Learn More
Hach Company Radiometer Analytical CDC741T Conductivity Cell with Temperature Sensor (2-pole Pt sensor, k = 1.0 cm-1, glass body)
Supplier: Hach Company E61M012
The Radiometer CDC741T is a 2-pole conductivity cell with platinized, platinum sensor (k = 1.0 cm-1), glass body and integrated temperature sensor. The CDC741T sensor's 2 platinum plate design is ideal for use with a sample changer due to easy rinsing. The cell has a fixed cable with MAB6 plug for use with Radiometer Analytical CDM210/230/92 conductivity meters. Please reference article no. E61M023 for a version of the CDC741T with RJ45 plug, compatible for use with Hach sensION+ benchtop conductivity meters.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.