Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Mouse anti-Human, Oligo, Clone: SK1, BD™ AbSeq ™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 940305
This item is not returnable.
View return policy
Description
CD8 Monoclonal antibody specifically detects CD8 in Human samples. It is validated for Single Cell 3' SequencingSpecifications
CD8 | |
Monoclonal | |
Oligo | |
CD8α; CD8A; CD8 alpha; Leu2a; MAL; T8; p32 | |
Human Peripheral Blood T Cells | |
RUO | |
Human | |
IgG1 κ |
Single Cell 3' Sequencing | |
Clone SK1 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction