Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD32b Monoclonal Antibody (A9C11), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA550622
Description
Positive control: Daudi cell lysate, Raji cell lysate, HL-60 cell lysate, human placenta tissue lysate, human tonsil tissue, human liver tissue, human colon carcinoma tissue, human placenta tissue. Predicted band size: 34 kDa Subcellular Location: Cell membrane.
This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.Specifications
CD32b | |
Monoclonal | |
1 mg/mL | |
PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P31994 | |
FCGR2B | |
Recombinant protein within human aa 1-250/310. | |
100 μL | |
Primary | |
Human | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
A9C11 | |
Unconjugated | |
FCGR2B | |
AI528646, CD32, CD32 receptor 2, CD32 receptor 2a, CD32 receptor 2b, CD32A, CD32B, CDw32, F630109E10Rik, Fc fragment of IgG receptor IIb, Fc fragment of IgG, low affinity II, receptor for (CD32), Fc fragment of IgG, low affinity IIb, receptor, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), fc gamma receptor IIB, Fc gamma RIIb, Fc receptor, IgG, low affinity IIb, Fc[g]RII, FCG2, FC-gamma RII, fc-gamma RII-a, Fc-gamma RII-b, Fc-gamma-RII, fc-gamma-RIIa, FcgammaRIIB, fc-gamma-RIIb, FcgammaRIIB2, FcGR, Fcgr2, Fcgr2a, FCGR2A1, FCGR2B, FcgRII, Fcr-2, Fcr-3, fcRII, fcRII-a, fcRII-b, FGFR2B, IGFR2, IgG Fc receptor II beta, igG Fc receptor II-b, Low affinity immunoglobulin gamma Fc region receptor II, low affinity immunoglobulin gamma Fc region receptor II-b, Ly-17, LyM-1, Ly-m20, Lymphocyte antigen 17 | |
Mouse | |
Protein A | |
RUO | |
2213 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG1 |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction