Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ CD FortiCHO™ Medium
![GREENER_CHOICE](/content/dam/fishersci/glyphs/greener_choice.png)
CD FortiCHO™ Medium
Supplier: Gibco™ A1148301
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Description
CD FortiCHO™ medium is specifically designed to offer the highest batch and fed-batch culture performance and yield with recombinant CHO cells in a chemically defined environment. The protein-free, animal origin component-free formulation provides consistency and helps reduce the need to screen for adventitious agents. With CD FortiCHO™ medium cell cultures last 2+ days longer than competitive alternatives, which means decreased HCP (Host Cell Protein) concentration for removal in downstream purification.CD FortiCHO™ medium is fortified to support maximal recombinant CHO cell growth and protein expression in suspension batch culture, offering 120% greater growth and production than select competitive media with simple glucose supplementation. CD FortiCHO™ medium can provide up to 75% greater fed-batch growth and production over glucose-only fed-batch culture for selected cell lines and applications when used with CD EfficientFeed™ C nutrient supplement.
When to Use CD FortiCHO™ Medium
When to Consider Another Gibco™ Medium
Consider CD OptiCHO™ medium when:
Consider CD CHO medium when:
Consider CD DG44 when:
Dry Media Formulations
For early access to CD FortiCHO™ AGT™ medium, please contact Gibco™ Technical Services.
Design Your Own Media Specific to Your Needs
Whether it’s a bench-level experiment or large scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs. With Gibco™ cGMP Media Custom Services you can put the manufacturing power and years of technical experience of our R&D and Bioproduction Application Specialists to work for you.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Storage: Store in the dark at +2 to +8°C. Guaranteed stable for 12 months when properly stored. |
|
Animal Origin-free, Chemically-defined, Protein-free, Serum-free | |
Suspension Cell Culture | |
CD OptiCHO AGT Medium | |
No Glutamine, No Phenol Red | |
Bottle | |
No antibiotics | |
1 L |
CHO | |
1 X | |
Liquid | |
Sodium Pyruvate, Sodium Bicarbonate | |
Low | |
Sustainable packaging | |
Gibco™ | |
Room Temperature |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?
Provide Content Correction