Learn More
Revvity Health Sciences Inc CCL2/MCP-1 (mouse) LANCE Ultra TR-FRET Detection Kit, 10,000 assay points
Supplier: Revvity Health Sciences Inc TRF1509M
The LANCEUltra Mouse CCL2/MCP-1? Detection Kit is designed for detection and quantitation of mouse Monocyte Chemoattractant Protein 1? in cell culture media using a homogeneous TR-FRET (no-wash steps, no separation steps) assay. The 500 point kit contains enough reagents to run 500 wells in 384-well format, using a 20 ¿L reaction volume (15 ¿L of sample). The 5,000 point kit contains enough reagents to run 5,000 wells in 384-well format, using a 20 ¿L reaction volume (15 ¿L of sample). No-wash steps, no separation steps TR-FRET technology Sensitive detection High reproducibility Faster time-to-results Easy automation 96-well, 384-well, and 1536-well formats LANCE and LANCE (Lanthanide chelate excite) Ultra are our TR-FRET (time-resolved fluorescence resonance energy transfer), homogeneous (no wash) technologies. One antibody of interest is labeled with a donor fluorophore (a LANCE Europium chelate) and the second molecule is labeled with an acceptor fluorophore (ULight dye). Upon exc
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.