Learn More
Foxx Life Sciences EZwaste HPLC Solvent Waste System, 10L Heavy Duty Reusable Carboy, 83mm (83B) VersaCap with Four (4x) Ports for 1/8 Inch O.D. Tubing; Three (3x) Ports for 1/4 Inch O.D. Tubing; One (1x) Hose Barb Port for 1/4 or 3/8 Inch I.D. Tubing
Supplier: Foxx Life Sciences 3323419OEM
332-3419-OEM: EZwaste System, 10L, HDPE, 83mm Cap, 4x 1/8", 3x 1/4" OD Tubing, 1/4" and 3/8" HB and Filter. Foxx Life Sciences EZwaste Solvent Waste System is designed to be an affordable solution for HPLC liquid waste disposal. Each system is manufactured to safely collect waste from two HPLC machines. This system features an activated carbon filter, which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers, and provides an easy mechanism of disposal for vapor wastes. The EZwaste Solvent Waste system also features a closed-loop design, which protects against leaks and spills, improving overall lab safety. Each EZwaste Solvent Waste System comes with one 83mm or 120mm open VersaCap, fittings, plugs for unused ports, a replaceable sulfur-cured EPDM gasket, and a chemical exhaust filter. * Depending on application, carbon exhaust filter life cycle may vary
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.