Learn More
World Precision Instrument High Temperature Cautery, 1500-2100F
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: World Precision Instrument 500388
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
For cutting, use the lower voltage with a continuous AC current to product heat rapidly and completely vaporize the tissue at the tip of the probe. For coagulation, use higher voltage in a pulsed manner to produce heat more slowly. With this method, tissue damage is more widespread (a few mm from the probe) and blood coagulates.
Note : For USA order only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.