Learn More
3B Scientific™ Catheterization simulator PRO
Used to realistically demonstrate, practice and evaluate female bladder catheterizations either via the urethra (transurethral) or the abdominal wall (suprapubic).
Supplier: 3B Scientific™ 1023009
Description
- The genital inserts are placed into the anatomically correct lower abdominal model and held in place with magnets. The material is soft and flexible: the labia are movable and can be separated.
- This way the student can practice all the necessary gestures for catheterization (e.g. disinfection). The material gives a realistic feeling when inserting and removing the catheter into the bladder with realistic resistance and insertion depths.
- Catheterization with realistic resistance
- Soft and movable (for practicing disinfection, for example)
- Anatomically realistic pelvic structure
- Liquid outflow if catheterization is successfully carried out
- The transparent bladder can be checked, and the abdominal wall can be removed
- Training of suprapubic catheter changes as well as their cleaning and care
- Stands securely on the table thanks to the non-slip feet
- Magnetic connectors for quick set-up and dismantling
- Easy to clean and maintain, and can be completely taken apart
- A sponge on the inside prevents a build-up of moisture
Specifications
Catheterization Trainer | |
To Train and Improve Competency in Male Catheterization. | |
Male |