Learn More
Enzo Life Sciences Catestatin (human) (1mg)
Supplier: Enzo Life Sciences BMLCP64200001
Human Chromogranin A [352-372]). Wild type catestatin, initially described as a potent catecholamine release inhibitory peptide. Cst located in the skin is active against Gram-negative and Gram-positive bacteria as well as against fungi and yeast. Also proposed as a modulator or regulator of cardiac function and as an anti-hypertensive agent. Alternative name: Chromogranin A (352-372). Purity: ≥98% (HPLC). Formulation: Lyophilized. Solubility: Soluble in Water. Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.