Learn More
Epredia™ Cassette 1 Slotted Tissue Cassettes
Hold tissue specimens during processing and embedding with Epredia™ Cassette 1 slotted tissue cassettes.
Supplier:  Epredia™ 1000963
 View more versions of this product
Catalog No. 10-009-63
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Acetal polymer construction
  • 30° writing face
  • Small slot-style pores
Ordering Alerts:
  • Supplied in box of 500 cassettes with attached breakaway lids (not available in Tube Packs)
Specifications
Specifications
Slotted Pore
Tan
Tissue Cassette
30°
Acetal Polymer