Learn More
Epredia™ Cassette 1 Slotted Tissue Cassettes
Hold tissue specimens during processing and embedding with Epredia™ Cassette 1 slotted tissue cassettes.
Supplier: Epredia™ 1000958
Description
- Acetal polymer construction
- 30° writing face
- Small slot-style pores
- Supplied in box of 500 cassettes with attached breakaway lids (not available in Tube Packs)
Specifications
Slotted Pore | |
Pink | |
Tissue Cassette |
30° | |
Acetal Polymer |