Learn More
Bioassay Systems QuantiFluo™ Caspase-3 Assay Kit. For quantitative determination of the apoptosis target caspase-3 activity and HTS screen for apoptosis inducers and inhibitors.
SDP
Supplier:  Bioassay Systems DCS3100
Encompass_Preferred
For quantitative determination of the apoptosis target caspase-3 activity and HTS screen for apoptosis inducers and inhibitors. Key Features: Safe. Non-radioactive assay. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: FL400/490nm. Samples: Cell and tissue lysate. Species: All. Procedure: Assay takes 60 min. Kit size: 100 tests.
Catalog No. 50-225-5860