Learn More
ProteinSimple Maurice CE-SDS Cartridge inserts, 2/pk
Supplier: ProteinSimple 046124
Cartridge Inserts for holding the top running buffer vial assembly (046-010) into the Maurice CE-SDS Cartridges (PS-MC02-S). 2 inserts/pk. For use with Maurice and Maurice S. systems only.