Learn More
Across International Stainless Steel Flask Carrier for Ai 20L Evaporating Flasks
Supplier: Across International SE.FC20L
Stainless steel 20L evaporating flask carrier with silicone cushions, custom made for Ai 20L SolventVap series rotary evaporators. This carrier can be placed on top of our 20L SolventVap's water bath for safe and easy flask loading and unloading; it can also serve as a flask table stand.