Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carolina™ Slime Mold Cultures
Slime mold is a broad term describing some organisms that use spores to reproduce.
Supplier: Carolina Biological Supply 156193
Alert:
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Description
- Myxomycetes
- Note: To ensure you order enough living materials for the number of students in your class, order one culture for up to 30 students, two cultures for 31 to 60 students, three cultures for 61 to 90 students, etc.
Specifications
Plate | |
Physarum polycephalum plasmodium | |
Class of 30 students | |
Bacteria Culture |
Ea. | |
Carolina™ techniques for studying bacteria and fungi manual | |
2% Agar with old fashioned Quaker™ oats | |
25°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction