Learn More
Carolina™ Individual Fungi Cultures
Fungi can help you teach many topics in ecology, genetics, biochemistry and other disciplines.
Supplier: Carolina Biological Supply 156250
Alert:
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Description
Fungi are important decomposers, helping recycle organic materials in food webs, which makes them great subjects for ecological studies. Students can culture them for genetic studies. For studies in biochemistry, students can investigate the role yeast plays in the production of wine, beer and baked goods.
- Available in tube cultures, plate cultures or MicroKwik Cultures™
- Labeled by name and media on which they are cultivated
- Guaranteed for purity and genera
- Incubate at room temperature (26°C)
Specifications
Teach many topics in ecology, genetics, biochemistry and other disciplines. | |
20°C to 22°C | |
Saccharomyces cerevisiae | |
Class of 30 students | |
Bacteria Culture |
Tube | |
Ea. | |
Carolina™ techniques for studying bacteria and fungi manual | |
Yeast Malt Agar | |
30°C |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.